Learn More
Description
- Filled in ISO 5 (Grade A/B, Former Class 100)
- Filtered at 0.2 microns
- Gamma irradiated to a sterility assurance level of 10-6
- Sterility tested according to current USP compendium
- Completely traceable from start to finish
- Completely validated for sterility and shelf life
Specifications
Specifications
| Product Type | Quaternary Ammonium Disinfectant |
| pH | Neutral |
| Sterility | Non-sterile |
| Certifications/Compliance | Certificate of Analysis |
| Container Type | Bottle |
| Filtered | Filtered |
| Format | SimpleMix |
| Volume (English) | 1 gal. |
| Volume (Metric) | 3.8 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.