Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decyltrimethoxysilane 97.0+%, TCI America™
SDP

Catalog No. D4704250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4704250G 250 g
D470425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4704250G Supplier TCI America Supplier No. D4704250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5575-48-4
Molecular Formula C13H30O3Si
Molecular Weight (g/mol) 262.47
MDL Number MFCD00069135
InChI Key KQAHMVLQCSALSX-UHFFFAOYSA-N
Synonym n-decyltrimethoxysilane, decyltrimethoxysilan, trimethoxydecylsilane, decyl trimethoxy silane, silane,decyltrimethoxy, decyl-trimethoxysilane
PubChem CID 2758023
IUPAC Name decyltrimethoxysilane
SMILES CCCCCCCCCC[Si](OC)(OC)OC

Specifications

Color Colorless
Boiling Point 106°C
Quantity 250 g
Formula Weight 262.47
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Decyltrimethoxysilane
EINECSNumber (2)-3512
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.