Learn More
Cayman Chemical Decylubiquinone, 55486-00-5, 10 mg
SDP
Supplier:  Cayman Chemical 2102710
Encompass_Preferred
Molecular Formula C19H30O4, Purity ≥98%, Formula Weight 322.4, Synonyms: 2-decyl-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione; An analog of ubiquinone that blocks opening of the mitochondrial permeability transition pore.
Catalog No. NC1291121
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.