Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Non-sterile Deep Well Plate for Storage and Collection

Catalog No. 10320177 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Round
Total Volume (Metric) Well:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Total Volume (Metric) Well
10-320-177 Round 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-320-177 Supplier Corning™ Supplier No. 3866

missing translation for 'validMainframeMsg'

Deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.

  • Made of polypropylene
  • Offer excellent chemical resistance
  • Can withstand temperatures down to -80°C
TRUSTED_SUSTAINABILITY

Specifications

Lid Without Lid
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Total Volume (Metric) Well 2 mL
Well Shape Round
Well Color Translucent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.