Learn More
Ampco Safety Tools Deep Well Impact Socket, 1/2 in. Drive (Metric)
SDP
Catalog No. 19173848 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
8 mm
9 mm
10 mm
11 mm
12 mm
13 mm
14 mm
15 mm
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
23 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
31 mm
32 mm
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-173-848 32 mm
19-173-812 10 mm
19-173-818 11 mm
19-173-819 12 mm
19-173-823 13 mm
19-173-824 14 mm
19-173-827 15 mm
19-173-828 16 mm
19-173-830 17 mm
19-173-831 18 mm
19-173-833 19 mm
19-173-834 20 mm
19-173-835 21 mm
19-173-836 22 mm
19-173-837 23 mm
19-173-838 24 mm
19-173-839 25 mm
19-173-840 26 mm
19-173-841 27 mm
19-173-842 28 mm
19-173-843 29 mm
19-173-846 30 mm
19-173-847 31 mm
19-173-852 8 mm
19-173-854 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-173-848 Supplier Ampco Safety Tools Supplier No. DWI1/2D32MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ DW Impact Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Deep-well Impact Socket Wrench
Drive Size 1/2 in.
Length (Metric) 32 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.