Learn More
Ampco Safety Tools Deep Well Impact Socket, 3/4 in. Drive (Metric)
SDP
Catalog No. 19173880 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
23 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
31 mm
32 mm
33 mm
34 mm
35 mm
36 mm
37 mm
38 mm
39 mm
40 mm
41 mm
42 mm
43 mm
46 mm
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-173-880 20 mm
19-173-875 17 mm
19-173-876 18 mm
19-173-878 19 mm
19-173-884 21 mm
19-173-885 22 mm
19-173-888 23 mm
19-173-889 24 mm
19-173-891 25 mm
19-173-892 26 mm
19-173-893 27 mm
19-173-894 28 mm
19-173-895 29 mm
19-173-897 30 mm
19-173-898 31 mm
19-173-899 32 mm
19-173-900 33 mm
19-173-901 34 mm
19-173-902 35 mm
19-173-903 36 mm
19-173-904 37 mm
19-173-905 38 mm
19-173-906 39 mm
19-173-907 40 mm
19-173-908 41 mm
19-173-909 42 mm
19-173-910 43 mm
19-173-911 46 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-173-880 Supplier Ampco Safety Tools Supplier No. DWI3/4D20MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ DW Impact Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Deep-well Impact Socket Wrench
Drive Size 3/4 in.
Length (Metric) 20 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.