Learn More
Mascon Medical Mascon Medical™ Deep Well Plate, 384-Well, 120ul, Sterile, 20EA/Bag, 5 Bag/Case
SDP
Supplier:  Mascon Medical FSDWP120
Encompass_Preferred
Mascon Medical™ 120ul Deep Well Plate is made of high-quality, medical-grade polypropylene with excellent chemical resistance. It is a reliable solution for sample storage, high-throughput screening, and low-binding experiments. It aligns with the international SBS standard and is suitable for automatic nucleic acid extraction and SBS automatic instruments. Best used for drug discovery, molecular biology, and genomic application. Works well with our Mascon Medical™ Sealing Film.Features:•Meets ISO13485 quality management system standard•Low retentionSpecifications:•Volume: 384-Well 120ul•Qty. / Case: 100•Color: Transparent•Shape: Square well, conical bottom•Material: Polypropylene•Sterile: Yes•Temperature Range: -80℃~120℃•Size: 127.3*85.3*14.3mm•Packaging Format: 20EA/Bag, 5Bags/Case
Catalog No. NC2458306
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.