Learn More
Ampco Safety Tools Deep Well Socket, 1/4 in. Drive (Imperial)
SDP
Catalog No. 19173654 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
1/2 in.
1/4 in.
11/32 in.
3/16 in.
3/8 in.
5/16 in.
7/16 in.
7/32 in.
9/16 in.
9/32 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-173-654 1/4 in.
19-173-653 1/2 in.
19-173-656 11/32 in.
19-173-661 3/16 in.
19-173-662 3/8 in.
19-173-666 5/16 in.
19-173-669 7/16 in.
19-173-670 7/32 in.
19-173-673 9/16 in.
19-173-674 9/32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-173-654 Supplier Ampco Safety Tools Supplier No. DW1/4D1/4
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Deep Well Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Deep-well Socket Wrench
Drive Size 1/4 in.
Length (English) 1/4 in.
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.