Learn More
Ampco Safety Tools Deep Well Socket, 1/4 in. Drive (Metric)
SDP
Catalog No. 19173672 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
4 mm
4.5 mm
5 mm
5.5 mm
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
13 mm
14 mm
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-173-672 8 mm
19-173-655 10 mm
19-173-657 11 mm
19-173-658 12 mm
19-173-659 13 mm
19-173-660 14 mm
19-173-663 4.5 mm
19-173-664 4 mm
19-173-665 5.5 mm
19-173-667 5 mm
19-173-668 6 mm
19-173-671 7 mm
19-173-675 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-173-672 Supplier Ampco Safety Tools Supplier No. DW1/4D8MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Deep Well Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Deep-well Socket Wrench
Drive Size 1/4 in.
Length (Metric) 8 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.