Learn More
Ampco Safety Tools Deep Well Socket, 3/4 in. Drive (Metric)
SDP
Catalog No. 19173737 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
19 mm
21 mm
22 mm
23 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
31 mm
32 mm
33 mm
34 mm
35 mm
36 mm
37 mm
38 mm
40 mm
41 mm
42 mm
46 mm
47 mm
50 mm
52 mm
54 mm
55 mm
56 mm
57 mm
58 mm
60 mm
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-173-737 19 mm
19-173-742 21 mm
19-173-743 22 mm
19-173-746 23 mm
19-173-747 24 mm
19-173-749 25 mm
19-173-750 26 mm
19-173-751 27 mm
19-173-752 28 mm
19-173-753 29 mm
19-173-755 30 mm
19-173-756 31 mm
19-173-757 32 mm
19-173-758 33 mm
19-173-759 34 mm
19-173-760 35 mm
19-173-761 36 mm
19-173-762 37 mm
19-173-763 38 mm
19-173-764 40 mm
19-173-765 41 mm
19-173-766 42 mm
19-173-767 46 mm
19-173-768 47 mm
19-173-769 50 mm
19-173-770 52 mm
19-173-771 54 mm
19-173-772 55 mm
19-173-773 56 mm
19-173-774 57 mm
19-173-775 58 mm
19-173-776 60 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-173-737 Supplier Ampco Safety Tools Supplier No. DW3/4D19MM
Only null left
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Deep Well Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Deep-well Socket Wrench
Drive Size 3/4 in.
Length (Metric) 19 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.