Learn More
Cayman Chemical DeethylIndnomycIn 1mg
SDP
Supplier:  Cayman Chemical 280891 MG
Encompass
An isoflavone glycoside scavenges 34 5 of generated DPPH radicals compared to a vehicle control in a cell-free assay at 100 g/ml inhibits LPS-induced cytotoxicity ROS production and cytoskeletal defects in HUVECs at 10 g/ml reduces brain infarct volume in a rat model of ischemia-reperfusion injury induced by MCAO at 26 8 mg/kg
Catalog No. 50-294-1804
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.