Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEFB119 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570887 Supplier Invitrogen™ Supplier No. PA570887
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 77%; Guinea Pig: 82%; Horse: 92%; Human: 100%; Pig: 92%; Rat: 85%.

This gene encodes a member of the beta subfamily of defensins. Beta-defensins are antimicrobial peptides that protect tissues and organs from infection by a variety of microorganisms. This gene is found in a cluster with other beta-defensin genes on the long arm of chromosome 20. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFB119
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DEFB119
Gene Accession No. Q8N690
Gene Alias Beta-defensin 119; beta-defensin 120; Beta-defensin 19; Beta-defensin 20; CBD119; CBD120; DEFB119; DEFB120; DEFB19; DEFB-19; DEFB20; DEFB-20; defensin beta 119; defensin, beta 119; Defensin, beta 120; defensin, beta 19; defensin, beta 20; ESC42-RELA; ESC42-RELB; UNQ2449/PRO5729
Gene Symbols DEFB119
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human DEFB119.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245932
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.