Learn More
Bachem rec b-Defensin 2 (human)
SDP
Supplier:  Bachem 4038287.0005
Encompass_Preferred
rec b-Defensin 2 (human), 479328-13-7, 5ug. rec hBD-2, rec BD-2 (human), rec Defensin b-2 (human), hBD-2. b-Defensin 2 (BD-2) (human), an antimicrobial peptide, is expressed throughout the epithelia of many organs. BD-2 is inducible upon contact of keratinocytes with Gram-negative bacteria or proinflammatory cytokines such as TNF-a and IL-1b. Elevated expression has been detected in lesional epidermal keratinocytes both in psoriasis and in the inflamed skin of mastitis.
Catalog No. 50-194-6673
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.