Learn More
TARGETMOL CHEMICALS INC DEFERASIROX 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T145750MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Deferasirox (CGP-72670) is an oral iron chelating agent used to treat chronic iron overload. purity: 99%
Catalog No. 50-257-7381
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.