Learn More
Apexbio Technology LLC Deferiprone, 1g Cas# 30652-11-0
SDP
Supplier:  Apexbio Technology LLC B17231000
Encompass_Preferred
Deferiprone is a chelating agent with an affinity for ferric ion (iron III), binds with ferric ions to form neutral 3:1 (deferiprone:iron) complexes that are stable over a wide range of pH values. 30652-11-0. MFCD00134497 Deferiprone is a chelating agent with an affinity for ferric ion (iron III)binds with ferric ions to form neutral 3:1 (deferiprone:iron) complexes that are stable over a wide range of pH values. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.