Learn More
Apexbio Technology LLC Deferiprone, 5g Cas# 30652-11-0
Supplier: Apexbio Technology LLC B17235000
Deferiprone is a chelating agent with an affinity for ferric ion (iron III), binds with ferric ions to form neutral 3:1 (deferiprone:iron) complexes that are stable over a wide range of pH values. 30652-11-0. MFCD00134497 Deferiprone is a chelating agent with an affinity for ferric ion (iron III)binds with ferric ions to form neutral 3:1 (deferiprone:iron) complexes that are stable over a wide range of pH values. There are other sizes available. Please inqury us for quote.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.