Learn More
Tocris Bioscience™ Deferoxamine mesylate
SDP
Catalog No. 5000176681
Encompass_Preferred
Catalog No. 50-001-76681 Supplier Tocris Bioscience™ Supplier No. HYP70390A5UG
Iron chelator; also hypoxia mimetic
Deferoxamine mesylate is an iron chelator. Also hypoxia mimetic and inhibitor of ferroptosis. Has neuroprotective effects in animal models of stroke, traumatic brain injury and neurodegenerative diseases.