Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Deferoxamine Mesylate, Iron Chelator, Solid, 1 g

Catalog No. 502584572
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4572 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4572 Supplier Abcam Supplier No. AB1207271G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Blood-brain barrier permeable iron chelator with immunomodulatory and anti-inflammatory properties.

  • Molecular Weight: 656.8 Da
  • Form: Solid
  • Function: Iron chelator; blood-brain barrier permeable
  • Biological Actions: Immunomodulation, inhibition of inflammation and atherosclerosis, and reduction of neuronal death
  • Storage Temperature: -20°C
  • Shipping Temperature: Ambient (can ship with ice)
  • CAS Number: 138-14-7
  • Citation Count: 14

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Iron
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Quantity 1 g
For Use With (Application) Immunomodulation, inhibition of inflammation, inhibition of atherosclerosis, reduction of neuronal death
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.