Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Deflagration Spoon
SCI_ED

Catalog No. S13828 Shop All GSC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
450 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S13828 450 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13828 Supplier GSC Go Science Crazy Supplier No. 43290110
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stainless Steel Deflecgation Spoon

The length and angle of the handle will potect your hands from coming in contact with a heat source. Made from Stainless Steel 8 in length, 3/4 in diameter bowl x 1/4 in depth of bowl.

Warranty and Services

1 year

Specifications

Material Stainless Steel
Length (Metric) 450 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.