Learn More
Abcam Deglycosylated Avidin, Native Chicken, >95% Purity, 100 μg
Description
- Purity: >95%
- Source: Native Chicken
- Tested Applications: ELISA, Flow Cyt, WB
- Formulation: Liquid, pH 7.3, PBS with 0.1% Sodium azide
- Storage: +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Avidin |
| Substrate Properties | Protein-based Substrate |
| Shipping Condition | Blue Ice |
| Content And Storage | Store at 4°C. |
| Form | Liquid |
| Buffer | PBS, 0.1% sodium azide, pH7.3 |
| Concentration | 0.1 mg/mL |
| Label or Dye | Unconjugated |
| Reactivity | Chicken |
| Formulation | PBS, pH 7.3, 0.1% Sodium azide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.