Learn More
Abcam Deglycosylated Avidin Protein, Native Chicken, HRP Conjugated, 1 mL
Description
- Protein Type: Native Chicken Full Length Deglycosylated Avidin protein
- Expression System: Native
- Purity: >90%
- Applications: WB, Flow Cyt, ELISA
- Formulation: Liquid, pH 7.4; contains 50% PBS and 49% Glycerol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 50% PBS, 49% glycerol, pH7.4 |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Label or Dye | HRP |
| Shipping Condition | Blue Ice |
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Biotin |
| Detection Method | Colorimetric |
| Formulation | 50% PBS, 49% Glycerol, pH 7.4 |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.