Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ degP Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144492 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144492 Supplier Invitrogen™ Supplier No. PA5144492
  • $412.50
    $372.65 / Each of 1

    Save $39.85

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

degP Polyclonal Antibody for Western Blot, ELISA

DegP acts as a chaperone at low temperatures but switches to a peptidase (heat shock protein) at higher temperatures (PubMed:10319814). Degrades transiently denatured and unfolded or misfolded proteins which accumulate in the periplasm following heat shock or other stress conditions (PubMed:16303867). DegP is efficient with Val-Xaa and Ile-Xaa peptide bonds, suggesting a preference for beta-branched side chain amino acids (PubMed:8830688). Only unfolded proteins devoid of disulfide bonds appear capable of being cleaved, thereby preventing non-specific proteolysis of folded proteins (PubMed:8830688). Its proteolytic activity is essential for the survival of cells at elevated temperatures (PubMed:7557477). It can degrade IciA, Ada, casein, globin and PapA. DegP shares specificity with DegQ (PubMed:8830688). DegP is also involved in the biogenesis of partially folded outer-membrane proteins (OMP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen degP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene degP
Gene Accession No. P0C0V0
Gene Alias htrA; ptd
Gene Symbols degP
Host Species Rabbit
Immunogen Recombinant Escherichia coli Periplasmic serine endoprotease DegP (degP) (27-474AA).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75202024
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.