Learn More
Essendant Heavy Duty Cleaner Degreaser Concentrate, 1 gal Bottle, 2/Carton
Supplier:  Essendant RAC89771CT
Encompass_Preferred
Heavy duty cleanser is specially formulated to cut through tough kitchen grease, fat and oil. This formula easily removes tough oily soil from a wide variety of surfaces and may be used in pressure washers and automatic scrubbers. This economical, concentrated formula makes up to 128 gallons (1:128). Use on counters, tables, grinders, refrigerators, concrete and ceramic floors, hoods and grills, engines, machinery, and tools.
Catalog No. 26-789-42
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.