Learn More
Abcam Zaprinast, cGMP phosphodiesterase inhibitor, 50MG
Supplier:  Abcam AB12094050MG
Encompass_Preferred
MW 271.28 g/mol, Purity >98%. Specific cGMP phosphodiesterase inhibitor, selective for PDE6, 5, 11 and 9 (IC₅₀ values are 0.15 (PDE6), 0.76 (PDE5), 12 (PDE11) and 29 μM (PDE9)). Reputed GPR35 agonist (EC₅₀ values are 16 and 840 nM at rat and human GPR35 receptors, respectively). Important mediator for a variety of cellular processes including vasodilation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7104
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.