Learn More
TARGETMOL CHEMICALS INC DDR INHIBITOR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T109855MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DDR inhibitor is a potent discidin domain receptor (DDR) inhibitor. The IC50 of DDR2 is 3.3 nM and DDR1 has 53% inhibition at 1.5 nM. purity: 97%
Catalog No. 50-258-1606
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.