Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEK Mouse anti-Human, Clone: 5E3A6, Proteintech
SDP

Catalog No. 501736628 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6628 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6628 Supplier Proteintech Group Inc Supplier No. 661941IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a protein with one SAP domain. This protein binds to cruciform and super helical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen DEK
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5E3A6
Concentration 1.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DEK
Gene Accession No. P35659
Gene Alias D6S231E, DEK, DEK oncogene, Protein DEK
Gene Symbols DEK
Host Species Mouse
Immunogen DEK Fusion Protein Ag8225
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7913
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.