Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Del-1, Mouse anti-Human, Clone: D345, Millipore Sigma™

Catalog No. p-7162962 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS197625 25 μg
MABS1976100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS197625 Supplier MilliporeSigma Supplier No. MABS197625UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detects Del-1 using this Anti-Del-1, clone D345, Cat. No. MABS1976, is a mouse monoclonal antibody that detects Developmentally-regulated endothelial cell locus 1 protein (Del-1) and has been tested for use in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Inhibition assay, & Western Blot.

EGF-like repeat and discoidin I-like domain-containing protein 3 (UniProt: O43854; also known as Developmentally-regulated endothelial cell locus 1 protein, Integrin-binding protein DEL1, Del-1) is encoded by the EDIL3 (also known as DEL1) gene (Gene ID: 10085) in human. Del-1 is a calcium-dependent glycoprotein that is secreted by endothelial cells and is shown to be expressed in a developmentally restricted pattern during embryogenesis. In adult tissues it is expressed in endothelial cells and in macrophages. Del-1 is reported to prevent apoptosis in endothelial cell subjected to TNF alpha, etoposide, and anoikis treatment. This anti-apoptotic effect is shown to be mediated specifically through binding of integrin alphav beta3 by the RGD motif. Del-1 expression is reported to be inversely related to that of IL-17 that is shown to inhibit Del-1 via a GSK-3beta and C/EBPb-dependent mechanism. This inhibitory action can be reversed at the PI3K/Akt signaling induced by D-resolvins. It serves as an endothelial cell-secreted homeostatic factor that regulates local inflammation in several tissues. It is reported to interfere with beta2-integrin-dependent inflammatory cell adhesion to endothelium and restricts transendothelial migration of neutrophils. Del-1 is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off in the mature form. It has three EGF-like domains in its N-terminal region. EGF-like domains 2 and 3 form a rigid rod via an interdomain calcium ion binding site, while the long linker between EGF1 and EGF2 lends considerable flexibility to EGF1. Del-1 also has discoidin I-like domains at the C-terminal region that mediate its binding to phosphatidylserine. Two isoforms of Del-1 have been described that are produced by alternative splicing. (Ref.: Dasgupta, SK et al. (2012). Circulation 125 (13); 1664-1672; Wang, Z., et al. (2012). Surgery 151(2); 296-305; Maekawa, T et al. (2015). Nat. Commun. DOI: 10.1038/ncomms9272).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Del-1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
Classification Monoclonal
Clone D345
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Del-1 in human cerebral cortex tissue sections.ELISA Analysis: A representative lot detected Del-1 in ELISA applications (Dasgupta, S.K., et. al. (2012). Ciculation. 125(13):1664-72; Maekawa, T., et. al. (2015). Nat Commun. 6:8272).Inhibition Analysis: A representative lot inhibited the uptake of BODIPY-labeled platelet microparticles by endothelium. (Dasgupta, S.K., et. al. (2012). Ciculation. 125(13):1664-72). Western Blotting Analysis: A representative lot detected Del-1 in Western Blotting applications (Dasgupta, S.K., et. al. (2012). Ciculation. 125(13):1664-72).Flow Cytometry Analysis: A representative lot detected Del-1 in Flow Cytometry applications (Dasgupta, S.K., et. al. (2012). Ciculation. 125(13):1664-72).
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias EGF-like repeat and discoidin I-like domain-containing protein 3;Developmentally-regulated endothelial cell locus 1 protein;Integrin-binding protein DEL1
Gene Symbols EDIL3;DEL1
Host Species Mouse
Immunogen GST-tagged full length human recombinant Developmentally-regulated endothelial cell locus 1 protein (Del-1).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_005702.3
Test Specificity Clone D345 detects human Developmentally-regulated endothelial cell locus 1 protein (Del-1).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.