Learn More
Cytiva T-series Centrasette cassette DELTA regenerated cellulose membrane 30 kDa MWCO 0 5 m
Supplier:  Cytiva DC030T06
Encompass
T-series Centrasette cassette DELTA regenerated cellulose membrane 30 kDa MWCO 0 5 m
Catalog No. 50-000-52875
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.