Learn More
CPC Scientific H-Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific DSIP001B
Encompass
A neuropeptide that when infused into the mesodiencephalic ventricle of recipient rabbits induces spindle and delta EEG activity and reduced motor activitiesONE-LETTER SEQUENCE: WAGGDASGEMOLECULAR FORMULA: C35H48N10O15MOLECULAR WEIGHT:848.82STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [62568-57-4], RESEARCH AREA: NeuropeptidesREFERENCES: G.A.Schoenenberger and M.Monnier, Proc. Natl. Acad. Sci. USA, 74, 1282 (1977)
Catalog No. 50-210-9200
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.