Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Delta-Tocotrienol, Vitamin E Form, Liquid, 1 mg

Catalog No. 502705711
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5711 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5711 Supplier Abcam Supplier No. AB1444491MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lipophilic antioxidant agent that induces apoptosis in cancer cells.

  • Molecular Weight: 396.6 Da
  • Purity: >98%
  • Form: Liquid
  • Properties: Lipophilic antioxidant agent; analog of tocotrienol α, β, and γ; induces apoptosis in cancer cells; enhances bone formation; shows antitumor, neuroprotective, and antioxidant effects in vivo; orally active
  • Storage Buffer: 99% Ethanol, 1% δ-tocotrienol
  • Storage Temperature: Store at -20°C
  • Shipping Temperature: Ambient

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Vitamin E (Delta-Tocotrienol)
Buffer 99% ethanol, 1% δ-tocotrienol
Content And Storage Store at -20°C.
Form Liquid
Product Type Antioxidant
Formulation 99% Ethanol, 1% δ-Tocotrienol
Quantity 1 mg
For Use With (Application) Antioxidant research, Cancer-apoptosis induction, Bone formation research, Neuroprotection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.