Learn More
delta-Valerolactone, 98%, may contain polymer
Description
δ-Valerolactone is employed as a chemical intermediate in the synthesis of polymers. It is also used to synthesize the copolyesters by lipase-catalyzed ring opening polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
δ-Valerolactone is employed as a chemical intermediate in the synthesis of polymers. It is also used to synthesize the copolyesters by lipase-catalyzed ring opening polymerization.
Solubility
Miscible with water.
Notes
Store in a tightly closed container. Avoid breathing dust, vapor, mist, or gas.
Specifications
Specifications
| Melting Point | -13°C to -12°C |
| Density | 1.056 |
| Boiling Point | 58°C to 60°C (0.5 mmHg) |
| Flash Point | 112°C (233°F) |
| Refractive Index | 1.458 |
| Quantity | 100 g |
| Beilstein | 106436 |
| Solubility Information | Miscible with water. |
| Formula Weight | 100.12 |
| Percent Purity | 98% |
| Show More |
Frequently Asked Questions (FAQs)
The quickest and simplest way to remove the polymeric material in the product is by filtering it through a pad of Celite under nitrogen.
Safety and Handling
| Hazard Category | H318 |
| Hazard Statement | GHS H Statement H318 Causes serious eye damage. |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| EINECSNumber | 208-807-1 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.