Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Deltamethrin, Protein Phosphatase 2B Inhibitor, Solid, 10 mg

Catalog No. 502706964
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6964 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6964 Supplier Abcam Supplier No. AB14101910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent protein phosphatase 2B inhibitor with over 98% purity, active in vivo.

  • Potent inhibitor: Protein phosphatase 2B (IC50 = 0.03 nM)
  • Mechanism: Induces neurotoxicity by slowing the opening and closing of Na+ channels
  • Purity: >98%
  • Molecular weight: 505.2 Da
  • Form: Solid
  • Active: In vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Form Solid
Buffer 100% deltamethrin
Target Molecule Protein Phosphatase 2B
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Formulation 100% Deltamethrin
Quantity 10 mg
For Use With (Application) Protein phosphatase 2B inhibition, Neurotoxicity research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.