Learn More
CPC Scientific H-Tyr-DMet-Phe-His-Leu-Met-Asp-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OPIO010A
Encompass
Deltorphin, YmFHLMD-amide, a peptide first isolated from skin extracts of the South American frog Phyllomedusa sauvagei, corresponds to the predicted prodermorphin heptapepeptide. Deltorphin acts as a highly potent and fully selective agonist for the delta-opioid receptor.ONE-LETTER SEQUENCE: Y-DMet-FHLMD-NH2MOLECULAR FORMULA: C44H62N10O10S2MOLECULAR WEIGHT:955.17STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [119975-64-3], RESEARCH AREA: PainREFERENCES: M.Amiche et al., Mol. Pharmacol., 35, 774 (1989); G.Kreil et al., Eur. J. Pharmacol., 162, 123 (1989); L.H.Lazarus et al., J. Biol. Chem., 264, 3047 (1989); A.Mor et al., FEBS Lett., 255, 269 (1989); S.Sagan et al., J. Biol. Chem., 264, 17100 (1989); P.Melchiorri et al., Eur. J. Pharmacol., 195, 201 (1991); S.Salvadori et al., J. Med. Chem., 34, 1656 (1991); L.H.Lazarus et al., J. Med. Chem., 34, 1350 (1991)
Catalog No. 50-210-9982
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.