Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Deluxe Burette Clamp, Single
SDPSCI_ED

Catalog No. S15891 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S15891 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15891 Supplier United Scientific™ Supplier No. CLBURDS
Add to Cart
Add to Cart

Sturdy Clamp that can be used with virtually all support stands and lattice setups.

  • Deluxe burette clamps are die-cast from aluminum.
  • Feature black rubber sleeves for firm gripping of burettes.
  • The adjustable clamps hold up to 30 mm O.D. burettes which are held in place by the pressure created by the springs.
  • For use with rods up to 19 mm diameter.
  • Available as a single or a double burette clamp

Specifications

Quantity 1 Ea.
Type Burette Clamp
Material Aluminum
Color Silver
Jaw Type Spring Loaded
Diameter (Metric) Inside 19 mm

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.