Learn More
United Scientific Supplies Deluxe Current Balance Kit
SDPSCI_ED
Catalog No. S27794 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Current Balance, Solenoid
Catalog No. Includes
S27794 Current Balance, Solenoid
1 options
Catalog No. S27794 Supplier United Scientific Supplies Supplier No. DCBA01KIT
Add to Cart
Add to Cart
Includes an air core solenoid and a current balance for demonstrating and measuring the small force experienced by a conductor carrying an electric current in a magnetic field
  • Solenoid's clear acrylic former and 90 mm diameter allow for easy viewing
  • Balance features an adjustable counterweight and can hold additional masses
  • Includes an activity guide
Specifications
Product Type Kit, Deluxe Current Balance
For Use With (Application) Demonstrating and Measuring The Small Force Experienced by a Conductor Carrying an Electric Current in a Magnetic Field
Includes Current Balance, Solenoid