Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Deluxe Double Burette Clamp
SDPSCI_ED

Catalog No. S82818 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Jaw Type:
Spring Loaded
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Jaw Type
S82818 Spring Loaded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning

Catalog No. S82818 Supplier United Scientific Supplies Supplier No. CLBURDD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning

Deluxe Burette Clamp offers a safe and sturdy way to hold two burettes.

  • Deluxe burette clamps are die-cast from aluminum.
  • Feature black rubber sleeves for firm gripping of burettes.
  • The adjustable clamps hold up to 30 mm O.D. burettes which are held in place by the pressure created by the springs.
  • For use with rods up to 19 mm diameter.

Specifications

Type Double Clamp
Material Cast Aluminum
Color Silver
Jaw Type Spring Loaded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.