Learn More
Description
- Lab coats made with heavier 1.75 oz spunbond polypropylene
- Provides non-hazardous splash and dry particulate protection
- Snap front
- Packaged 25 per case
- Three total pockets, one breast and two hip
- White, knit cuffs, knit collar
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | Large |
| Gender | Unisex |
| Material | Spunbond Polypropylene |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Single-use |
| Sterility | Non-sterile |
| No. of Pockets | 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.