Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Deluxe Metalware Kit
SDPSCI_ED

Catalog No. S111037 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Height (Metric):
Various
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
S111037 Various
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S111037 Supplier United Scientific™ Supplier No. UNMTLK1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Deluxe Metalware Kit includes everything you need to get your lab station ready to go.

This deluxe set includes:

  • Support Stand (5 x 8 in.) with Support Rod (20 in.)
  • Support Ring (3 in.)
  • 2-prong Extension Clamp with Cork Lined Jaws and a Clamp Holder
  • The support rod includes a 3 in. long rod to provide more torque when securing the rod to the base plate.

Specifications

Material Cast Iron
Height (Metric) Various
Diameter (English) Inner 3 in.
Diameter (Metric) Inner 76.2 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.