Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Deluxe Micrometer
SDPSCI_ED

Catalog No. S139596 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S139596 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Prop 65
Catalog No. S139596 Supplier United Scientific™ Supplier No. MCRDLX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Prop 65

Accurate and simple to use micrometer.

  • Micrometer is made of hardened steel for precision and durability
  • Range: 0 to 25 mm
  • Precision rotary scale has a resolution of 0.01 mm
  • Ratchet thimble and locking lever
  • Instructions are included

Specifications

Grade 5+
Includes Micrometer
Class Size 1 Lab Group
Product Type Micrometer

Warning: Cancer and Reproductive Harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.