Learn More
United Scientific™ Deluxe Slotted Weight Set of 8, with Rack
SDPSCI_ED
Catalog No. S139460 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
Set of 8
Catalog No. Quantity
S139460 Set of 8
1 options
This item is not returnable. View return policy
Catalog No. S139460 Supplier United Scientific™ Supplier No. WHST08
Add to Cart
Add to Cart
This item is not returnable. View return policy
High quality weights that last long in the classroom.
Set of 8 high-quality slotted brass weights with durable rack.
Weights included:
  • 1 x 500 g
  • 2 x 200 g
  • 1 x 100 g
  • 1 x 50 g
  • 2 x 20 g
  • 1 x 10 g
Specifications
Product Type Weight Set
Material Brass
Weight (Metric) 10, 20, 50, 100, 200, 500 g
Quantity Set of 8
Warning: Cancer and reproductive harm
Prop 65