Learn More
Market Lab Deluxe Titan Draw Cart
Supplier:  Market Lab 34843
Encompass_Preferred
The Deluxe Titan Draw Cart by MarketLab is a durable stainless steel cart with 5 drawers on wheels for medical and lab use. It has a weight capacity of 500 lbs and measures 41.5" H x 23.5" W x 22" D. The drawers have full extension slides for easy access. This mobile cart is made of 20 gauge stainless steel with a brushed finish. It comes fully assembled and has 5" medical grade casters with brakes.
Catalog No. NC2595096
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.