Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DEM1 Recombinant Protein Antigen
SDP

Catalog No. NB815971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB815971 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB815971 Supplier Novus Biologicals™ Supplier No. NBP188493PEP
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 64789
Molecular Weight (g/mol) 27 kDa
Quantity 0.1 mL
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias C1orf176, chromosome 1 open reading frame 176, defects in morphology 1 homolog (S. cerevisiae), Defects in morphology protein 1 homolog, EC 3.1.11.-, Exo V, FLJ11445, FLJ13183, FLJ21144
Gene Symbol EXO5
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-88493. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL.
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein DEM1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.