Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Demecolcine, ≥98%, MP Biomedicals™

Catalog No. ICN19019510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19019510 10 mg
ICN19019501 1 mg
ICN19019505 5 mg
ICN19019550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19019510 Supplier MP Biomedicals, Inc Supplier No. 0219019510
Add to Cart
Add to Cart

Chemical Identifiers

CAS 477-30-5
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00075459
InChI Key NNJPGOLRFBJNIW-UHFFFAOYNA-N
Synonym demecolcine, colcemid, colchamine, --demecolcine, reichstein's f, colcemide, demecolcin, desmecolcine, santavy's substance f, --colchamine
PubChem CID 220401
ChEBI CHEBI:4393
SMILES CNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12

Specifications

Color Yellow
Assay Percent Range >98%
Quantity 10 mg
IUPAC Name (7S)-1,2,3,10-tetramethoxy-7-(methylamino)-6,7-dihydro-5H-benzo[a]heptalen-9-one
Formula Weight 371.4
Physical Form Powder
Chemical Name or Material Demecolcine, Antineoplastic agent
Recommended Storage Store at + 4°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.