Description
Demecolcine has been used as a microtubule depolymerizing agent. It has also been used to inhibit mitotic division.
Specifications
Specifications
| Quantity | 5 mg |
| Label Type | Other Labels or Dyes |
| Product Type | Stain |