Learn More
Demecolcine, Sigma-Aldrich
Catalog No. 01259029 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-029 10 mg
01-258-859 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-029 Supplier MilliporeSigma Supelco Supplier No. D738510MG
Add to Cart
Add to Cart
Demecolcine has been used as a microtubule depolymerizing agent. It has also been used to inhibit mitotic division.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Label Type Other Labels or Dyes
Product Type Stain