Learn More
Apexbio Technology LLC Demethylzeylasteral 107316-88-1 20mg
SDP
Supplier:  Apexbio Technology LLC N276320
Encompass_Preferred
Demethylzeylasteral (CAS 107316-88-1) is a triterpenoid small molecule isolated from the whole plant or peeled wood of Tripterygium wilfordii Hook f It is known for modulating immune responses particularly by inhibiting the activation and proliferation of T lymphocytes and downregulating pro-inflammatory cytokine production Through these immunosuppressive activities demethylzeylasteral is investigated in models of autoimmune diseases and inflammation The compound is of interest for studying the molecular pathways underlying immune regulation and is commonly utilized in preclinical pharmacology research related to immunomodulation Store in a sealed cool and dry environment
Catalog No. 50-000-94235
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.