Learn More
Thermo Scientific™ Demineralizer Cartridges

Description
Hose nipples on each end of these cartridges easily connect to your water supply with ordinary lab tubing (0.3 in. I.D. [9.5mm]). Resins in the deionization cartridge contain a special dye that changes color when the cartridge is exhausted.
High-Capacity Cartridge
- Has a two-bed demineralizer with selected cation and anion resin beds
- Delivers more liters per cartridge than mixed-bed types
- Average resistance of effluent is 175kilohm·cm
- Total ion-exchange capacity is 1725 grains (as NaCl)
- Produces high-resistance demineralized water from tap water
- Increases purity of water from central demineralizer
- Total ion-exchange capacity is 1200 grains (as NaCl), 849 grains (as NaCl) to 1 megohm·cm endpoint
- pH is usually between 6.8 and 7.2
- Water is largely free of silica and CO2
Specifications
Specifications
| Capacity (English) | 1200 grains as NaCl; 849 grains as NaCl |
| Type | Cartridge, Ultrapure Demineralizer |
| For Use With (Application) | Water purification |
| Diameter (Metric) | 9.5 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.