Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Demonstration Balance Support
SDPSCI_ED

Catalog No. S13060 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13060 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13060 Supplier United Scientific Supplies Supplier No. DBS002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

18.5cm high

Sturdy support for meter stick demonstration balances and torque experiments. The cast metal support is 18.5cm high. Requires knife-edge clamps, slotted weights and hangers, and a meter stick.

Specifications

Grade 6 to 12
Format Kit
Product Type Demonstration Balance
Height (Metric) 18.5 cm
Languages English
For Use With (Application) Knife Edged Lever clamp, meter stick, and weights
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.