Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Demonstration Motor/Generator Set
SDPSCI_ED

Catalog No. S07366 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Activity guide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S07366 Activity guide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07366 Supplier United Scientific™ Supplier No. DMG002
Add to Cart
Add to Cart

Use this device as either a motor or generator demonstrator in your classroom.

  • Works as a motor and uses electrical energy to perform mechanical work
  • As a generator, it converts mechanical work to electrical energy
  • Includes an activity guide

Specifications

For Use With (Application) Used as either a motor or generator demonstration in classroom by converting mechanical work to electrical energy
Includes Activity guide
Product Type Demonstration Motor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.