Learn More
United Scientific™ Demonstration Pulleys
SDPSCI_ED
Catalog No. S16027 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Double Parallel Pulley
Single Pulley
Triple Parallel Pulley
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S16027 Double Parallel Pulley
S15683 Single Pulley
S15688 Triple Parallel Pulley
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S16027 Supplier United Scientific™ Supplier No. PUDD01
Add to Cart
Add to Cart
Ideal for use in demonstrating simple machines.
  • This jumbo-sized double pulley is designed for classroom demonstrations.
  • Measures 70 mm in diameter, with an open type design for easy operation
  • Lightweight, low-friction, and deformation-resistant plastic ensures accurate results
  • Available in single, double, or triple pulleys.
Specifications
Product Type Double Parallel Pulley
Diameter (Metric) 70 mm
Warning: Cancer and reproductive harm